CH6371379500 Chem Impex International, Inc.

L-a-tert-Butylglycine-tert-butyl ester hydrochloride

  • MDL number MFCD00038904

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-alpha-tert-Butyl-Gly-OtBu HCl
Purity 95
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-alpha-i-tert-i-butylglycine-i-tert-i-butyl-ester-hydrochloride/4090?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-alpha-i-tert-i-butylglycine-i-tert-i-butyl-ester-hydrochloride/4090?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart