CH6371379501 Chem Impex International, Inc.

O-tert-Butyl-L-threonine allyl ester hydrochloride

  • MDL number MFCD00798676
  • Cas Number : 218938-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Thr(tBu)-OAll HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-l-threonine-allyl-ester-hydrochloride/4091?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-l-threonine-allyl-ester-hydrochloride/4091?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart