CH6371379503 Chem Impex International, Inc.

L-Threonine methylamide

  • MDL number MFCD00238186
  • Cas Number : 79009-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Thr-NHMe
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-threonine-methylamide/4096?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-threonine-methylamide/4096?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart