CH6371379505 Chem Impex International, Inc.

L-Threonine b-naphthylamide

  • MDL number MFCD00080957
  • Cas Number : 729-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-142� C
Synonym L-Thr-betaNA
Purity 99
Storage temp. Store at RT
Physical Form Off -white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-threonine-ß-naphthylamide/4098?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-threonine-ß-naphthylamide/4098?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart