CH6371379506 Chem Impex International, Inc.

Nin-Boc-L-tryptophan tert-butyl ester hydrochloride

  • MDL number MFCD00798682
  • Cas Number : 218938-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-160 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-in-sup-boc-l-tryptophan-i-tert-i-butyl-ester-hydrochloride/4099?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-in-sup-boc-l-tryptophan-i-tert-i-butyl-ester-hydrochloride/4099?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart