CH6371379508 Chem Impex International, Inc.

O-tert-Butyl-L-tyrosine allyl ester hydrochloride

  • MDL number MFCD00798675
  • Cas Number : 218938-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Tyr(tBu)-OAll HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-l-tyrosine-allyl-ester-hydrochloride/4102?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-l-tyrosine-allyl-ester-hydrochloride/4102?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart