CH6371379511 Chem Impex International, Inc.

L-Valine amide hydrobromide

  • MDL number MFCD00050704
  • Cas Number : 33529-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-valine-amide-hydrobromide/4112?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-valine-amide-hydrobromide/4112?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart