CH6371379514 Chem Impex International, Inc.

L-Valine 4-nitrobenzyl ester hydrobromide

  • MDL number MFCD00058245
  • Cas Number : 6015-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-158
Synonym L-Val-O(4-NO2Bzl) HBr
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-valine-4-nitrobenzyl-ester-hydrobromide/4115?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-valine-4-nitrobenzyl-ester-hydrobromide/4115?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart