CH6371379516 Chem Impex International, Inc.

L-a-Aminoadipic acid d-methyl ester hydrochloride

  • MDL number MFCD01632070
  • Cas Number : 147780-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Aad(OMe)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-alpha-aminoadipic-acid-delta-methyl-ester-hydrochloride/4120?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-alpha-aminoadipic-acid-delta-methyl-ester-hydrochloride/4120?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart