CH6371379526 Chem Impex International, Inc.

3-N-Maleimidobenzoic acid

  • MDL number MFCD00022572
  • Cas Number : 17057-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Light yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-i-n-i-maleimidobenzoic-acid/4130?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-i-n-i-maleimidobenzoic-acid/4130?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart