CH6371379536 Chem Impex International, Inc.

Na-Boc-Nb-allyloxycarbonyl-L-2,3-diaminopropionic acid

  • MDL number MFCD00798611
  • Cas Number : 161561-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-92
Synonym Boc-L-Dap(Aloc)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-beta-sup-allyloxycarbonyl-l-2-3-diaminopropionic-acid/4140?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-beta-sup-allyloxycarbonyl-l-2-3-diaminopropionic-acid/4140?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart