CH6371379537 Chem Impex International, Inc.

Nalpha-Boc-Nbeta-bromoacetyl-L-2,3-diaminopropionic acid

  • MDL number MFCD00798614
  • Cas Number : 135630-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-ß-sup-bromoacetyl-l-2-3-diaminopropionic-acid/4141?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-ß-sup-bromoacetyl-l-2-3-diaminopropionic-acid/4141?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart