CH6371379545 Chem Impex International, Inc.

Na-Fmoc-Ng-(1-{1'-adamantyl}-1-methyl-ethoxycarbonyl)-L-2,4-diaminobutyric acid

  • MDL number MFCD01632073
  • Cas Number : 214750-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Dab(Adpoc)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-sup-1-1-adamantyl-1-methyl-ethoxycarbonyl-l-2-4-diaminobutyric-acid/4149?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-sup-1-1-adamantyl-1-methyl-ethoxycarbonyl-l-2-4-diaminobutyric-acid/4149?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart