CH6371379549 Chem Impex International, Inc.

Na,g-Bis-Boc-D-2,4-diaminobutyric acid dicyclohexylammonium salt

  • MDL number MFCD01632075

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Dab(Boc)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-bis-boc-d-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/4153?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-bis-boc-d-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/4153?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart