CH6371379551 Chem Impex International, Inc.

Na,g-Bis-Z-L-2,4-diaminobutyric acid

  • MDL number MFCD00236965
  • Cas Number : 55478-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Dab(Z)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-bis-z-l-2-4-diaminobutyric-acid/4155?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-bis-z-l-2-4-diaminobutyric-acid/4155?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart