CH6371379555 Chem Impex International, Inc.

Na-Z-L-2,4-diaminobutyric acid

  • MDL number MFCD00077904
  • Cas Number : 62234-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182 - 190
Synonym Z-L-Dab-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-2-4-diaminobutyric-acid/4159?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-2-4-diaminobutyric-acid/4159?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart