CH6371379557 Chem Impex International, Inc.

Na-Z-Ng-Boc-L-2,4-diaminobutyric acid dicyclohexylammonium salt

  • MDL number MFCD00237341
  • Cas Number : 3350-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-201
Synonym Z-L-Dab(Boc)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-gamma-sup-boc-l-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/4161?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-gamma-sup-boc-l-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/4161?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart