CH6371379559 Chem Impex International, Inc.

Ng-Boc-L-2,4-diaminobutyric acid methyl ester hydrochloride

  • MDL number MFCD00798680
  • Cas Number : 3350-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-164
Synonym L-Dab(Boc)-OMe HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-gamma-sup-boc-l-2-4-diaminobutyric-acid-methyl-ester-hydrochloride/4163?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-gamma-sup-boc-l-2-4-diaminobutyric-acid-methyl-ester-hydrochloride/4163?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart