CH6371379560 Chem Impex International, Inc.

N-1-Trityl-deamino-histidine

  • MDL number MFCD00237290
  • Cas Number : 160446-35-5
  • Molecular Weight : 382.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at = -4 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-i-n-i-1-trityl-imidazol-4-yl-propionic-acid/4164?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-i-n-i-1-trityl-imidazol-4-yl-propionic-acid/4164?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart