CH6371379561 Chem Impex International, Inc.

N-Methyl-alpha-aminoisobutyric acid hydrochloride

  • MDL number MFCD06795786
  • Cas Number : 2566-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-197 °C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-alpha-aminoisobutyric-acid-hydrochloride/4165?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-alpha-aminoisobutyric-acid-hydrochloride/4165?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart