CH6371379562 Chem Impex International, Inc.

N-Methyl-L-aspartic acid b-tert-butyl ester

  • MDL number MFCD00672353
  • Cas Number : 197632-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Me-L-Asp(OtBu)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-aspartic-acid-ß-i-tert-i-butyl-ester/4167?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-aspartic-acid-ß-i-tert-i-butyl-ester/4167?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart