CH6371379572 Chem Impex International, Inc.

Na-Methyl-L-histidine hydrochloride

  • MDL number MFCD00077143
  • Cas Number : 17451-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Me-L-His-OH HCl
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-methyl-l-histidine-hydrochloride/4177?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-methyl-l-histidine-hydrochloride/4177?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart