CH6371379573 Chem Impex International, Inc.

Nalpha-Methyl-L-histidine methyl ester hydrochloride

  • MDL number MFCD00672362
  • Cas Number : 118384-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at =-4°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-methyl-l-histidine-methyl-ester-hydrochloride/4178?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-methyl-l-histidine-methyl-ester-hydrochloride/4178?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart