CH6371379574 Chem Impex International, Inc.

N-Methyl-L-Leucine benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00083659
  • Cas Number : 42807-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-134
Synonym N-Me-L-Leu-OBzl TosOH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-leucine-benzyl-ester-4-toluenesulfonate-salt/4179?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-leucine-benzyl-ester-4-toluenesulfonate-salt/4179?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart