CH6371379576 Chem Impex International, Inc.

N-Methyl-O-methyl-L-tyrosine hydrochloride

  • MDL number MFCD06795806
  • Cas Number : 52939-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Me-L-Tyr(Me)-OH HCl
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-methyl-i-o-i-methyl-l-tyrosine-hydrochloride/4182?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-methyl-i-o-i-methyl-l-tyrosine-hydrochloride/4182?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart