CH6371379577 Chem Impex International, Inc.

N-Methyl-4-nitro-L-phenylalanine

  • MDL number MFCD00038839
  • Cas Number : 70663-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to light yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-4-nitro-l-phenylalanine/4183?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-4-nitro-l-phenylalanine/4183?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart