CH6371379579 Chem Impex International, Inc.

Nalpha-Methyl-L-phenylalanine methyl ester hydrochloride

  • MDL number MFCD00083661
  • Cas Number : 19460-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-methyl-l-phenylalanine-methyl-ester-hydrochloride/4185?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-methyl-l-phenylalanine-methyl-ester-hydrochloride/4185?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart