CH6371379585 Chem Impex International, Inc.

Sarcosine amide hydrochloride

  • MDL number MFCD00058283
  • Cas Number : 5325-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Sar-NH2 HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/sarcosine-amide-hydrochloride/4194?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/sarcosine-amide-hydrochloride/4194?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart