CH6371379587 Chem Impex International, Inc.

Sarcosine benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00191015
  • Cas Number : 54384-06-4
  • Molecular Weight : 351.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at = -4 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/sarcosine-benzyl-ester-4-toluenesulfonate-salt/4196?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/sarcosine-benzyl-ester-4-toluenesulfonate-salt/4196?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart