CH6371379588 Chem Impex International, Inc.

Z-g-carboxy-g-(di-tert-butyl ester)-D-glutamic acid

  • MDL number MFCD00038757

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-D-Gla(OtBu)2-OH
Purity 99
Storage temp. Store at = -4
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-gamma-carboxy-gamma-di-i-tert-i-butyl-ester-d-glutamic-acid/4197?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-gamma-carboxy-gamma-di-i-tert-i-butyl-ester-d-glutamic-acid/4197?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart