CH6371379589 Chem Impex International, Inc.

Z-gamma-carboxy-gamma-(di-tert-butyl ester)-DL-glutamic acid

  • MDL number MFCD00066103
  • Cas Number : 56877-43-1
  • Molecular Weight : 437.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at = -4 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-gamma-carboxy-gamma-di-i-tert-i-butyl-ester-dl-glutamic-acid/4198?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-gamma-carboxy-gamma-di-i-tert-i-butyl-ester-dl-glutamic-acid/4198?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart