CH6371379591 Chem Impex International, Inc.

Z-3-(1-naphthyl)-L-alanine

  • MDL number MFCD00238419
  • Cas Number : 65365-15-3
  • Molecular Weight : 349.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-168 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-3-1-naphthyl-l-alanine/4200?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-3-1-naphthyl-l-alanine/4200?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart