CH6371379592 Chem Impex International, Inc.

Z-3-(2-naphthyl)-L-alanine

  • MDL number MFCD00063338
  • Cas Number : 65365-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-114
Synonym Z-L-Ala(2-naphthyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-3-2-naphthyl-l-alanine/4202?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-3-2-naphthyl-l-alanine/4202?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart