CH6371379593 Chem Impex International, Inc.

Z-3-aminobenzoic acid N-hydroxysuccinimide ester

  • MDL number MFCD00238395
  • Cas Number : 129666-48-4
  • Molecular Weight : 368.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-149 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-3-aminobenzoic-acid-i-n-i-hydroxysuccinimide-ester/4203?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-3-aminobenzoic-acid-i-n-i-hydroxysuccinimide-ester/4203?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart