CH6371379596 Chem Impex International, Inc.

Z-Ala-Met-OH

  • MDL number MFCD00056133
  • Cas Number : 76264-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 119 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-ala-met-oh/4207?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-ala-met-oh/4207?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart