CH6371379597 Chem Impex International, Inc.

Z-Ala-Phe-OH

  • MDL number MFCD00020425
  • Cas Number : 2768-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-alanyl-L-phenylalanine
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-ala-phe-oh/4208?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-ala-phe-oh/4208?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart