CH6371379599 Chem Impex International, Inc.

Na-Z-Nw-(4-methoxybenzenesulfonyl)-L-arginine dicyclohexylammonium salt

  • MDL number MFCD00237326
  • Cas Number : 58810-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Arg(Mbs)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-i-n-i-sup-sup-4-methoxybenzenesulfonyl-l-arginine-dicyclohexylammonium-salt/4210?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-i-n-i-sup-sup-4-methoxybenzenesulfonyl-l-arginine-dicyclohexylammonium-salt/4210?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart