CH6371379601 Chem Impex International, Inc.

Nalpha-Z-L-asparagine tert-butyl ester

  • MDL number MFCD00056253
  • Cas Number : 25456-85-3
  • Molecular Weight : 322.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 - 107 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-l-asparagine-i-tert-i-butyl-ester/4215?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-l-asparagine-i-tert-i-butyl-ester/4215?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart