CH6371379603 Chem Impex International, Inc.

Z-L-aspartic acid b-N-hydroxysuccinimide ester a-methyl ester

  • MDL number MFCD00237340
  • Cas Number : 54743-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Asp(OSu)-OMe
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-ß-i-n-i-hydroxysuccinimide-ester-a-methyl-ester/4220?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-ß-i-n-i-hydroxysuccinimide-ester-a-methyl-ester/4220?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart