CH6371379607 Chem Impex International, Inc.

Z-3-(1-naphthyl)-D-alanine

  • MDL number MFCD00672544
  • Cas Number : 96402-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-152 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-3-1-naphthyl-d-alanine/4227?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-3-1-naphthyl-d-alanine/4227?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart