CH6371379611 Chem Impex International, Inc.

Na-Z-Nw-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-D-arginine cyclohexylammonium salt

  • MDL number MFCD00237328
  • Cas Number : 200191-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-D-Arg(Pbf)-OH CHA
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-omega-sup-2-2-4-6-7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine-cyclohexylammonium-salt/4231?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-omega-sup-2-2-4-6-7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine-cyclohexylammonium-salt/4231?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart