CH6371379612 Chem Impex International, Inc.

Na-Z-D-arginine hydrochloride

  • MDL number MFCD00063010
  • Cas Number : 113712-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-D-Arg-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-d-arginine-hydrochloride/4233?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-d-arginine-hydrochloride/4233?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart