CH6371379615 Chem Impex International, Inc.

Na-Z-D-histidine

  • MDL number MFCD00067702
  • Cas Number : 67424-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-D-His-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-d-histidine/4240?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-d-histidine/4240?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart