CH6371379617 Chem Impex International, Inc.

Na,e-Z-L-Bis-D-lysine

  • MDL number MFCD00065212
  • Cas Number : 69677-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-D-Lys(Z)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-sup-a-e-sup-z-l-bis-d-lysine/4242?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-sup-a-e-sup-z-l-bis-d-lysine/4242?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart