CH6371379618 Chem Impex International, Inc.

Nalpha-Z-D-lysine benzyl ester benzenesulfonate salt

  • MDL number MFCD00672347
  • Cas Number : 201018-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-d-lysine-benzyl-ester-benzenesulfonate-salt/4243?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-d-lysine-benzyl-ester-benzenesulfonate-salt/4243?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart