CH6371379619 Chem Impex International, Inc.

Z-b-tert-butyl-D-alanine�A

  • MDL number MFCD02259501
  • Cas Number : 201677-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-154
Synonym Z-D-beta-tBu-Ala-OH DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-beta-i-tert-i-butyl-d-alanine-font-size-2-face-symbol-middot-font-dcha/4244?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-beta-i-tert-i-butyl-d-alanine-font-size-2-face-symbol-middot-font-dcha/4244?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart