CH6371379622 Chem Impex International, Inc.

Na-Z-D-ornithine

  • MDL number MFCD00237357
  • Cas Number : 112229-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-D-Orn-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-d-ornithine/4248?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-d-ornithine/4248?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart