CH6371379623 Chem Impex International, Inc.

Z-D-phenylalanine N-hydroxysuccinimide ester

  • MDL number MFCD00672535
  • Cas Number : 3397-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-140 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-phenylalanine-i-n-i-hydroxysuccinimide-ester/4249?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-phenylalanine-i-n-i-hydroxysuccinimide-ester/4249?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart