CH6371379626 Chem Impex International, Inc.

Z-D-serine benzyl ester

  • MDL number MFCD00237364
  • Cas Number : 53933-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81 - 87 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-serine-benzyl-ester/4254?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-serine-benzyl-ester/4254?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart