CH6371379627 Chem Impex International, Inc.

Z-DL-Ala-Gly-OH

  • MDL number MFCD00021731
  • Cas Number : 2503-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-dl-ala-gly-oh/4256?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-dl-ala-gly-oh/4256?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart